Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-Crown 5-Ether 97.0+%, TCI America™
SDP

Catalog No. C085925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C085925ML 25 mL
C08595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C085925ML Supplier TCI America Supplier No. C085925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33100-27-5
Molecular Formula C10H20O5
Molecular Weight (g/mol) 220.265
MDL Number MFCD00005110
InChI Key VFTFKUDGYRBSAL-UHFFFAOYSA-N
Synonym 15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
PubChem CID 36336
ChEBI CHEBI:32401
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadecane
SMILES C1COCCOCCOCCOCCO1

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 116°C
Quantity 25 mL
Formula Weight 220.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 15-Crown 5-Ether
RTECSNumber SB0200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.