Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-Lipoxygenase 1 Antibody (OTI3G8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP270169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270169 Supplier Novus Biologicals Supplier No. NBP270169
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

15-Lipoxygenase 1 Monoclonal specifically detects 15-Lipoxygenase 1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 15-Lipoxygenase 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3G8
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias 15-lipooxygenase-1, 15-LOX-1, 15LOX-1, arachidonate 15-lipoxygenase, Arachidonate omega-6 lipoxygenase, EC 1.13.11, EC 1.13.11.33, LOG15,15-LOX
Gene Symbols ALOX15
Host Species Mouse
Immunogen Full length human recombinant protein of human ALOX15(NP_001131) produced in HEK293T cell.
Molecular Weight of Antigen 74.6 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 246
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Rat, Canine, Monkey, Primate
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.