Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-Lipoxygenase 1 Antibody (OTI3G8), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270169FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270169FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270169FR Supplier Novus Biologicals Supplier No. NBP270169FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

15-Lipoxygenase 1 Monoclonal specifically detects 15-Lipoxygenase 1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 15-Lipoxygenase 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3G8
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias 15-lipooxygenase-1, 15-LOX-1, 15LOX-1, arachidonate 15-lipoxygenase, Arachidonate omega-6 lipoxygenase, EC 1.13.11, EC 1.13.11.33, LOG15,15-LOX
Gene Symbols ALOX15
Host Species Mouse
Immunogen Full length recombinant protein of human 15-Lipoxygenase 1 (NP_001131) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 246.0
Target Species Human, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.