Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-PGDH/HPGD Antibody (004), PE, Novus Biologicals™
SDP

Catalog No. NB317462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317462 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317462 Supplier Novus Biologicals Supplier No. NBP289838PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen 15-PGDH/HPGD
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 004
Conjugate PE
Dilution ELISA, Sandwich ELISA Capture
Formulation PBS
Gene Alias 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human 15-PGDH/HPGD (Accession#: NP_000851.2; Met1-Gln266).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3248
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.