Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17-Beta-Estradiol Antibody (4S112 (BGN/06/88112)), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10062279R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062279R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062279R Supplier Novus Biologicals Supplier No. NB10062279R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen 17-Beta-Estradiol
Applications ELISA
Classification Monoclonal
Clone 4S112 (BGN/06/88112)
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD
Host Species Mouse
Immunogen 17-beta estradiol conjugated to bovine serum albumin at position 6.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62279 is specific for 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. NB100-62279 exhibits negligible cross reactivity with estriol and testosterone.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.