Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17-Beta-Estradiol Antibody, Novus Biologicals™
SDP

Catalog No. NB10062282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062282 0.25 mL
NB017455 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062282 Supplier Novus Biologicals Supplier No. NB10062282
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

17-Beta-Estradiol Polyclonal specifically detects 17-Beta-Estradiol in Non-species specific samples. It is validated for ELISA, Radioimmune Assays (RIA).

Specifications

Antigen 17-Beta-Estradiol
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:5000-1:40000
Formulation PBS with 0.09% Sodium Azide
Gene Alias 17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD
Host Species Sheep
Immunogen 17-beta estradiol conjugated to bovine serum albumin at position 6.
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62282 recognizes 17 beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Oestradiol 100% Oestrone 14% Oestriol 5% Aldosterone, Cortisol, Corticosterone, DHEA, Deoxycorticosterone, Pregnenolone, Progesterone, 17OH-P and Testosterone all <0.01%
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.