Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
17-Beta-Estradiol Antibody, Novus Biologicals™

Sheep Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | 17-Beta-Estradiol |
---|---|
Concentration | 5.0 mg/mL |
Dilution | ELISA 1:5000-1:40000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017455
![]() |
Novus Biologicals
NB100622820.05ML |
0.05 mL |
Each for $206.50
|
|
|||||
NB10062282
![]() |
Novus Biologicals
NB10062282 |
0.25 mL |
Each for $499.50
|
|
|||||
Description
17-Beta-Estradiol Polyclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.Specifications
17-Beta-Estradiol | |
ELISA 1:5000-1:40000 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.09% Sodium Azide | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5.0 mg/mL | |
ELISA | |
Unconjugated | |
Sheep | |
All species | |
IgG | |
Protein G purified | |
NB100-62282 recognizes 17 beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Oestradiol 100% Oestrone 14% Oestriol 5% Aldosterone, Cortisol, Corticosterone, DHEA, Deoxycorticosterone, Pregnenolone, Progesterone, 17OH-P and Testosterone all <0.01% |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title