Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17 beta-HSD1/HSD17B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15629520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15629520 20 μL
NBP156295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15629520 Supplier Novus Biologicals Supplier No. NBP15629520UL

Rabbit Polyclonal Antibody has been used in 2 publications

17 beta-HSD1/HSD17B1 Polyclonal specifically detects 17 beta-HSD1/HSD17B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen 17 beta-HSD1/HSD17B1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml, Immunohistochemistry 4-8 ug/ml, Immunohistochemistry-Paraffin 4-8 ug/ml
Formulation From PBS. with No Preservative
Gene Accession No. P14061
Gene Alias 17-beta-HSD 1, 17-beta-hydroxysteroid dehydrogenase type 1, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E17KSR, E2DH, EC 1.1.1.62, EDH17B1, EDH17B2EDHB17, estradiol 17-beta-dehydrogenase 1, estradiol 17-beta-dehydrogenase-1, HSD17, hydroxysteroid (17-beta) dehydrogenase 1, hydroxysteroid (17-beta) dehydrogenase 1 isoform, MGC138140, Placental 17-beta-hydroxysteroid dehydrogenase, SDR28C1, short chain dehydrogenase/reductase family 28CE, member 1
Gene Symbols HSD17B1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to HSD17B1(hydroxysteroid (17-beta) dehydrogenase 1) The peptide sequence was selected from the N terminal of HSD17B1 (NP_000404). Peptide sequence MARTVVLITGCSSGIGLHLAVRLASDPSQSFKVYATLRDLKTQGRLWEAA.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3292
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.