Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17 beta-HSD14/HSD17B14 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB080587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080587 Supplier Novus Biologicals Supplier No. NBP296996C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

17 beta-HSD14/HSD17B14 Polyclonal specifically detects 17 beta-HSD14/HSD17B14 in Human samples. It is validated for ELISA.

Specifications

Antigen 17 beta-HSD14/HSD17B14
Applications ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant 17 beta-HSD14/HSD17B14 (NP_057330.2; Met1-Ser270).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51171
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.