Learn More
Description
Includes
- Detector Industrial Kit includes detector with monochrome display, integral pump, 10
- ft. sampling line and 1 ft. probe. Detector Deluxe Kit includes detector with color display, integral pump, 10 ft. sampling line and 1 ft. probe
New XCell™ sensor technology doubles the typical life span of sensors, offers faster response and span calibration times and provides plug-and-play capabilities for easy reconfiguration. Sensors available for SO2, Cl2 and NH3 for expanded monitoring applications.
- Sensor response and clear times under 15 seconds for most common sensor configurations
- End of sensor life warning provides advanced notice to user, reducing service outages
- MotionAlert activates if user becomes disabled and motionless and provides user's location
- InstantAlert enables users to manually alert others to potentially hazardous situations
- Span calibration time of 60 seconds for most common sensor configurations
- 24-hour bump checkmark; bump test under 15 seconds for most common configurations
- Robust integral pump for consistent flow without externally attached components
- 18 language options
- Durable, rubberized housing for secure grip
- Lithium-ion battery lasts up to 20 hours
- High-contrast monochrome or color display
- Fully compatible with Galaxy™ Automated Test System and MSA Link™ software
Order Info
L x W x D: 6.69 x 3.49 x 1.79 in.. Weight: 1 lb.
Compliance
IP65 rated. Approvals: Class I, Division 1, Groups A, B, C and D; Class II Division 1 Groups E, F and G; Class III Division 1
Warranty and Services
Three years on detector, CO, H2S, O2, LEL and SO2 sensors.
Specifications
Specifications
| Alarm Types | Audible, visual, vibrating |
| Battery Life | 20 hr. |
| Components Measured | LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide |
| Dimensions (L x W x H) | 3.5 x 3.8 x 3.75 in. (8.89 x 9.65 x 9.53cm) |
| Type | Multigas Detector: Standard Kit |
| Height (English) | 3.75 in. |
| Height (Metric) | 9.53 cm |
| IP Rating | IP65 |
| Length (English) | 3.5 in. |
| Length (Metric) | 8.89 cm |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
