Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18-Crown-6, 99%

Catalog No. AC181565000
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle

CAS: 17455-13-9 | C12H24O6 | 264.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17455-13-9
Molecular Formula C12H24O6
Molecular Weight (g/mol) 264.32
MDL Number MFCD00005113
InChI Key XEZNGIUYQVAUSS-UHFFFAOYSA-N
Synonym 18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
PubChem CID 28557
ChEBI CHEBI:32397
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCO1

Specifications

Melting Point 36.0°C to 40.0°C
Color Colorless or White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 500 g
Fieser 04,142; 05,152; 06,133; 07,76; 08,1280; 09,126; 12,403; 15,76; 17,290
Merck Index 14,2602
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hydrocarbons,benzene,chloroform,methanol and trichlorotrifluoroethane
Formula Weight 264.32
Percent Purity 99%
Physical Form Crystalline Mass, Crystals or Liquid
Chemical Name or Material 18-Crown-6
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 241-473-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.