Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Benzimidazole-2-methanol, 97%, Thermo Scientific™

Catalog No. AC304190050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4856-97-7
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00014560
InChI Key IAJLTMBBAVVMQO-UHFFFAOYSA-N
Synonym 1h-benzimidazole-2-methanol, 1h-benzoimidazol-2-yl methanol, 2-benzimidazolemethanol, 2-hydroxymethyl benzimidazole, 1h-benzo d imidazol-2-yl methanol, 2-hydroxymethylbenzimidazole, 2-hydroxymethyl-1h-benzimidazole, 1h-benzoimidazol-2-yl-methanol, 1h-1,3-benzodiazol-2-ylmethanol, 1h-benzimidazol-2-yl methanol
PubChem CID 78569
IUPAC Name 1H-benzimidazol-2-ylmethanol
SMILES C1=CC=C2C(=C1)NC(=N2)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 170°C to 175°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Benzimidazole-2-methanol, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-451-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.