Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-1-Amino-2-(diphenylphosphino)indane, 97+%, Thermo Scientific™

Catalog No. p-7041001
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6093103 1 g
AAH6093106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6093103 Supplier Thermo Scientific Chemicals Supplier No. H6093103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1091606-70-0
Molecular Formula C21H20NP
Molecular Weight (g/mol) 317.372
MDL Number MFCD17013983
InChI Key IALYUOSURKZWLR-NHCUHLMSSA-N
Synonym 1r,2r-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine, 1r,2r-2-diphenylphosphanyl-2,3-dihydro-1h-inden-1-amine, 1r,2r-1-amino-2-diphenylphosphino indane, 1r,2r-1-amino-2-diphenylphosphino indane, 97+%, 1r,2r-2-diphenylphosphino indan-1-amine, 1r,2r-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine hydrochloride, 1r,2r-2-diphenylphosphino-2,3-dihydro-1h-inden-1-amine,min 10wt% in thf
PubChem CID 59444846
IUPAC Name (1R,2R)-2-diphenylphosphanyl-2,3-dihydro-1H-inden-1-amine
SMILES C1C(C(C2=CC=CC=C21)N)P(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Quantity 1 g
Formula Weight 317.36
Percent Purity ≥97%
Chemical Name or Material (1R,2R)-1-Amino-2-(diphenylphosphino)indane
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.