Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 1R,3R-RSL3

Click to view available options
Quantity:
10 mg
50 mg
Description
1R,3R-RSL3 is a negative control for 1S,3R-RSL3 (Cat. No. 6118).
Specifications
Specifications
| Chemical Name or Material | (1 R,3 R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1 H-pyrido[3,4-b]indole-3-carboxylate |
| CAS | 1219810-15-7 |
| Quantity | 10 mg |
| Target | Glutathione Peroxidase 4/GPX4 |
| Molecular Formula | C23H21ClN2O5 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction