Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,3R)-ACPD, Tocris Bioscience™

Catalog No. p-4783088
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-841-0 10 mg
02-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-841-0 Supplier R&D Systems Supplier No. 0284/10
This item is not returnable. View return policy

Group I and group II mGlu agonist; active isomer of (±)-trans-ACPD (Cat. No. 0187)

Active isomer of (±)-trans-ACPD . Agonist at both group I and II mGlu receptors (EC50 values are 5, 15, 42 and 60 μM at mGluR2, mGluR5, mGluR1 and mGluR6 respectively).

NPEC-caged-(1S,3R)-ACPD, (±)-trans-ACPD and cis-ACPD also available.

Chemical Identifiers

CAS 111900-32-4
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
InChI Key YFYNOWXBIBKGHB-FBCQKBJTSA-N
Synonym 1s,3r-1-aminocyclopentane-1,3-dicarboxylic acid, 1s,3r-acpd, trans-acpd, 1-amino-1,3-dicarboxycyclopentane, trans-+/--acpd, chembl34453, 1s,3s-acpd, 1-aminocyclopentane-1s,3r-dicarboxylic acid, 1s,3r-1-acpd
PubChem CID 104766
IUPAC Name (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
SMILES C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 173.17
Percent Purity >98%
Chemical Name or Material (1S, 3R)-ACPD
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.