Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 1S,3R-RSL3
GREENER_CHOICE

Catalog No. p-7116100
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)

1S,3R-RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor. Induces ferroptosis. Increases lipid peroxidase levels in vitro. Inhibits tumor formation and progression in vivo.

Negative control 1R,3R-RSL3 (Cat. No. 6687) also available.

Specifications

Chemical Name or Material (1S,3R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylate
CAS 1219810-16-8
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H21ClN2O5
Formula Weight Observed MW: 440.88
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.