Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(2-Chloroethoxy)ethoxy]ethanol, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5197-62-6
C6H13ClO3
168.62
MFCD00002874
KECMLGZOQMJIBM-UHFFFAOYSA-N
2-2-2-chloroethoxy ethoxy ethanol,ethanol, 2-2-2-chloroethoxy ethoxy,triethylene glycol monochloride,triethylene glycol monochlorohydrin,ethanol,2-2-2-chloroethoxy ethoxy,2-2-2-chloro-ethoxy ethoxy ethanol,2-2-2-chloroethoxy ethoxy-ethanol
78871
2-[2-(2-chloroethoxy)ethoxy]ethanol
C(COCCOCCCl)O
5197-62-6
C6H13ClO3
168.62
MFCD00002874
KECMLGZOQMJIBM-UHFFFAOYSA-N
2-2-2-chloroethoxy ethoxy ethanol,ethanol, 2-2-2-chloroethoxy ethoxy,triethylene glycol monochloride,triethylene glycol monochlorohydrin,ethanol,2-2-2-chloroethoxy ethoxy,2-2-2-chloro-ethoxy ethoxy ethanol,2-2-2-chloroethoxy ethoxy-ethanol
78871
2-[2-(2-chloroethoxy)ethoxy]ethanol
C(COCCOCCCl)O
Specific Gravity 1.163
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 256.0°C to 258.0°C
Quantity 50g
Density 1.1630g/mL
Color Yellow
Flash Point 107°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula Cl(CH2)2O(CH2)2OCH2OH
Beilstein 01, 468
Formula Weight 168.62
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-[2-(2-Chloroethoxy)ethoxy]ethanol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-984-