Learn More
2,2,2-Tribromoethanol, 99%
Description
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.
Solubility
Soluble in alcohol, ether, benzene and warmpetroleum ether. Partially soluble in water.
Notes
Light Sensitive. Store in cool and well ventilated area. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 77°C to 80°C |
| Boiling Point | 92°C to 93°C (10 mmHg) |
| Quantity | 5 g |
| Beilstein | 1733249 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in alcohol,ether,benzene and warmpetroleum ether. Partially soluble in water. |
| Formula Weight | 282.76 |
| Percent Purity | 99% |
| Chemical Name or Material | 2,2,2-Tribromoethanol |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 200-903-1 |
| RTECSNumber | KM3675000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.