Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxycarbonyl isocyanate, 90%, tech.

Catalog No. AC314310050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 19617-43-7 | C4H5NO3 | 115.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19617-43-7
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
InChI Key VMVZGGPZNHFGKS-UHFFFAOYSA-N
Synonym ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester
PubChem CID 300166
IUPAC Name ethyl N-(oxomethylidene)carbamate
SMILES CCOC(=O)N=C=O

Specifications

Color Colorless
Density 1.1100g/mL
Boiling Point 25.0°C (10.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4090
Linear Formula C2H5O2CNCO
Quantity 5 g
Specific Gravity 1.11
Formula Weight 115.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethoxycarbonyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.