Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3-Trimethylbutane 96.0+%, TCI America™
SDP

Catalog No. T071125ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Catalog No. T071125ML

Supplier: TCI America T071125ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 464-06-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00039846
InChI Key ZISSAWUMDACLOM-UHFFFAOYSA-N
Synonym triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
PubChem CID 10044
IUPAC Name 2,2,3-trimethylbutane
SMILES CC(C)C(C)(C)C

Specifications

Melting Point -24°C
Color Colorless
Boiling Point 81°C
Quantity 25 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3-Trimethylbutane
EINECSNumber (2)-0007
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.