Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98%

Catalog No. ac156250500
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 4390-04-9 | C16H34 | 226.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4390-04-9
Molecular Formula C16H34
Molecular Weight (g/mol) 226.44
MDL Number MFCD00008856
InChI Key VCLJODPNBNEBKW-UHFFFAOYNA-N
Synonym isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
PubChem CID 20414
IUPAC Name 2,2,4,4,6,8,8-heptamethylnonane
SMILES CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7900g/mL
Boiling Point 240.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Linear Formula (CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3
Quantity 50 mL
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Formula Weight 226.44
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
I
EINECSNumber 224-506-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.