Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4-Tetramethylpentane, 98%

Catalog No. AAB2164209
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAB2164209

Supplier: Thermo Scientific Chemicals B2164209

Only null left
Add to Cart

CAS: 1070-87-7 | C9H20 | 128.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-87-7
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00060873
InChI Key GUMULFRCHLJNDY-UHFFFAOYSA-N
PubChem CID 14058
IUPAC Name 2,2,4,4-tetramethylpentane
SMILES CC(C)(C)CC(C)(C)C

Specifications

Density 0.72
Boiling Point 121°C
Flash Point 6°C (43°F)
Odor Petroleum-like
Linear Formula (CH3)3CCH2C(CH3)3
Quantity 10 g
UN Number UN3295
Synonym di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl
Formula Weight 128.26
Percent Purity 98%
Chemical Name or Material 2,2,4,4-Tetramethylpentane
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.