Learn More
2,2,4-Trimethylpentane, ACS, 99+%
Description
Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -107°C |
| Color | Colorless |
| Density | 0.692 g/cm3 at 20°C |
| Boiling Point | 98°C to 99°C |
| Flash Point | -7°C (19°F) |
| Odor | Nearly Odorless |
| Assay Percent Range | >99% |
| Refractive Index | 1.391 |
| Quantity | 1 L |
| UN Number | 1262 |
| Show More |
Safety and Handling
| Hazard Category | H225-H304-H315-H319-H336 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 3; Packing Group: II |
| EINECSNumber | 208-759-1 |
| RTECSNumber | SA3320000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.