2,2,4-Trimethylpentane, ACS, >99%, Alfa Aesar™

Manufacturer: Alfa Aesar 31787AP

Quantity 500mL

 View more versions of this product

Catalog No. AA31787AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 540-84-1
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula (CH3)2CHCH2C(CH3)3
    Formula Weight 114.23
    MDL Number MFCD00008943
    Synonym Isooctane
    Chemical Name or Material 2, 2, 4-Trimethylpentane
    Grade ACS Grade

    Specifications

    Formula Weight 114.23
    Quantity 500mL
    UN Number 1262
    Boiling Point 98 to 99°C
    Melting Point &minus107°C
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 208-759-1
    Physical Form Liquid
    Odor Nearly Odorless
    Vapor Pressure 15 hPa (11mm Hg) at 20°C
    Assay Percent Range >99%
    Molecular Formula (CH3)2CHCH2C(CH3)3
    MDL Number MFCD00008943
    CAS 540-84-1
    Synonym Isooctane
    Appearance Colorless liquid
    Health Hazard 1 H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.
    Chemical Name or Material 2, 2, 4-Trimethylpentane
    Grade ACS Grade

    H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit