Learn More
2,2,4-Trimethylpentane, Environmental Grade, 99.5+%
Description
Suitable for pesticide residue analysis2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -107°C |
| Density | 0.692 g/mL |
| Boiling Point | 98°C to 99°C |
| Flash Point | -7°C (19°F) |
| Odor | Nearly odorless |
| Refractive Index | 1.391 |
| Quantity | 4 x 4 L |
| UN Number | UN1262 |
| Beilstein | 1696876 |
| Merck Index | 14,5193 |
| Show More |
Safety and Handling
| Hazard Category | H225-H304-H315-H319-H336 |
| Hazard Statement | GHS H Statement H225-H304-H315-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES |
| EINECSNumber | 208-759-1 |
| RTECSNumber | SA3320000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.