Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Aminoethoxy)ethanol, 98%

Catalog No. p-4509356
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 929-06-6 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-06-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
InChI Key GIAFURWZWWWBQT-UHFFFAOYSA-N
Synonym 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine
PubChem CID 13578
IUPAC Name 2-(2-aminoethoxy)ethanol
SMILES C(COCCO)N

Specifications

Melting Point -12.5°C
Color Yellow
Density 1.0600g/mL
Boiling Point 218.0°C to 224.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4585 to 1.4605
Linear Formula H2NCH2CH2OCH2CH2OH
Quantity 100 mL
Beilstein 04,III,642
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and aromatic hydrocarbons, practically immiscible with aliphatic hydrocarbons, and ethyl ether
Formula Weight 105.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-(2-Aminoethoxy)ethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 213-195-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.