Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2', Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis, MP Biomedicals™

Catalog No. ICN19502301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19502301 1 g
ICN19502305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19502301 Supplier MP Biomedicals, Inc Supplier No. 0219502301

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Quantity 1 g
Formula Weight 548.7
Physical Form Powder
Chemical Name or Material 2, 2' Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.