Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyrimidyl 95.0+%, TCI America™
SDP

Catalog No. B24961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24961G 1 g
B2496200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24961G Supplier TCI America Supplier No. B24961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34671-83-5
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00014600
InChI Key HKOAFLAGUQUJQG-UHFFFAOYSA-N
Synonym 2,2'-bipyrimidine, 2,2'-bipyrimidyl, 2,2-bipyrimidine, bpym, 2-pyrimidin-2-yl pyrimidine, bipyrimidine, 2,2'-bipyrimidinyl, 2,2'-dipyrimidyl, 2,2;-bipyrimidine
PubChem CID 123444
IUPAC Name 2,2'-bipyrimidine
SMILES C1=CN=C(N=C1)C1=NC=CC=N1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 158.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyrimidyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.