Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Butoxyethoxy)ethanol, 99+%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
112-34-5
C8H18O3
162.23
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
2-2-butoxyethoxy ethanol,diethylene glycol monobutyl ether,butyldiglycol,butyl carbitol,diethylene glycol butyl ether,butyl diglycol,butyl digol,ethanol, 2-2-butoxyethoxy,butoxyethoxyethanol,butoxydiglycol
8177
2-(2-butoxyethoxy)ethanol
CCCCOCCOCCO
112-34-5
C8H18O3
162.23
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
2-2-butoxyethoxy ethanol,diethylene glycol monobutyl ether,butyldiglycol,butyl carbitol,diethylene glycol butyl ether,butyl diglycol,butyl digol,ethanol, 2-2-butoxyethoxy,butoxyethoxyethanol,butoxydiglycol
8177
2-(2-butoxyethoxy)ethanol
CCCCOCCOCCO
Viscosity 6.16 mPa.s (20°C)
Flash Point 100°C
Quantity 1L
Specific Gravity 0.955
Density 0.9550g/mL
Boiling Point 231.0°C
Refractive Index 1.4310 to 1.4330
Melting Point -68.0°C
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)3OCH2CH2OCH2CH2OH
Beilstein 01,IV,2394
Merck Index 15,3133
Solubility Information Solubility in water: soluble. Other solubilities: miscible with oils, miscible with most common organic solvents
Formula Weight 162.23
Percent Purity 99+%
Chemical Name or Material 2-(2-Butoxyethoxy)ethanol, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.