Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Chloroethoxy)ethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
628-89-7
C4H9ClO2
124.57
MFCD00002870
LECMBPWEOVZHKN-UHFFFAOYSA-N
2-2-chloroethoxy ethanol,ethanol, 2-2-chloroethoxy,diglycol chlorohydrin,diglycol chlorhydrin,2-chloroethyl 2-hydroxyethyl ether,monochloropolyoxyethylene,2-2-chloroethoxy ethan-1-ol,unii-r2up818l4i,diethylene glycol monochlorohydrin,polyethylene glycol chloride 210
12361
2-(2-chloroethoxy)ethanol
C(COCCCl)O
628-89-7
C4H9ClO2
124.57
MFCD00002870
LECMBPWEOVZHKN-UHFFFAOYSA-N
2-2-chloroethoxy ethanol,ethanol, 2-2-chloroethoxy,diglycol chlorohydrin,diglycol chlorhydrin,2-chloroethyl 2-hydroxyethyl ether,monochloropolyoxyethylene,2-2-chloroethoxy ethan-1-ol,unii-r2up818l4i,diethylene glycol monochlorohydrin,polyethylene glycol chloride 210
12361
2-(2-chloroethoxy)ethanol
C(COCCCl)O
Specific Gravity 1.18
Flash Point 90°C
Density 1.1800g/mL
Boiling Point 180°C to 185°C
Water 1% max. (K.F.)
Infrared Spectrum Authentic
Refractive Index 1.4519 to 1.4539
Color Undesignated
Packaging Glass bottle
Quantity 50mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula ClCH2CH2OCH2CH2OH
Beilstein 01, 467
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with ethyl alcohol and diethyl ether
Formula Weight 124.57
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-(2-Chloroethoxy)ethanol, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 211-059-9

RTECSNumber : KK1350000

TSCA : TSCA