Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Cyanophenoxy)acetamide, 98%

Catalog No. AAH6627106
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 54802-12-9 | C9H8N2O2 | 176.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54802-12-9
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00297091
InChI Key RYOYNMJXEHNYAG-UHFFFAOYSA-N
Synonym 2-2-cyanophenoxy acetamide, acetamide, 2-2-cyanophenoxy, 2-cyanophenoxyacetamide
PubChem CID 12353988
IUPAC Name 2-(2-cyanophenoxy)acetamide
SMILES C1=CC=C(C(=C1)C#N)OCC(=O)N

Specifications

Quantity 5 g
UN Number UN3439
Beilstein 2939357
Formula Weight 176.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(2-Cyanophenoxy)acetamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.