Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
656-46-2
C8H4F2O4
202.12
VJLDRFCNFNQTTH-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid,2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid,2,2-difluorobenzodioxole-5-carboxylic acid,2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid,3,4-difluoromethylenedioxy benzoic acid,acmc-209nrl,5-carboxy-2,2-difluoro-1,3-benzodioxole,2,2-difluorobenzo-1,3dioxole-5-carboxylic acid,2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid,2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
608772
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
656-46-2
C8H4F2O4
202.12
VJLDRFCNFNQTTH-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid,2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid,2,2-difluorobenzodioxole-5-carboxylic acid,2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid,3,4-difluoromethylenedioxy benzoic acid,acmc-209nrl,5-carboxy-2,2-difluoro-1,3-benzodioxole,2,2-difluorobenzo-1,3dioxole-5-carboxylic acid,2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid,2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
608772
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
Melting Point 153°C to 157°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Formula Weight 202.12
Percent Purity 97%
Chemical Name or Material 2, 2-Difluoro-1, 3-benzodioxole-5-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning