Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-4-pentenoic acid, 95%

Catalog No. aah6094003
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH6094003

Supplier: Thermo Scientific Chemicals H6094003

Only null left
Add to Cart

CAS: 16386-93-9 | C7H12O2 | 128.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16386-93-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00012304
InChI Key BGUAPYRHJPWVEM-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g
PubChem CID 140065
IUPAC Name 2,2-dimethylpent-4-enoic acid
SMILES CC(C)(CC=C)C(O)=O

Specifications

Density 0.933
Boiling Point 104°C to 108°C
Flash Point 88°C (190°F)
Refractive Index 1.431
Linear Formula H2C=CHCH2C(CH3)2CO2H
Quantity 1 g
UN Number UN3265
Solubility Information Soluble in water.
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material 2,2-Dimethyl-4-pentenoic acid
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.