Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Ethoxyethoxy)ethanol, Spectrum™ Chemical
SDP

Catalog No. 18608143 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
200 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-143 200 L Glass Bottle
18-608-146 500 mL Glass Bottle
18-608-142 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-143 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1265200LTBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 111-90-0

Specifications

CAS Max % 1
Boiling Point 196°C to 197°C (384.8°F to 386.6°F)
Packaging Glass Bottle
Quantity 200 L
Molecular Weight (g/mol) 134.18
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy)ethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.