Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-(Ethylenedioxy)bis(ethylamine), 97+%

Catalog No. AA4475930
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 929-59-9 | C6H16N2O2 | 148.206 g/mol

2,2'-(Ethylenedioxy)bis(ethylamine) is used to linker to form conjugates of manganese-doped zinc sulfide nanoparticles with folic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-59-9
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.206
MDL Number MFCD00040474
InChI Key IWBOPFCKHIJFMS-UHFFFAOYSA-N
Synonym 1,8-diamino-3,6-dioxaoctane, 1,2-bis 2-aminoethoxy ethane, 2,2'-ethylenedioxy bis ethylamine, unii-r41zt4uf34, 2,2'-ethane-1,2-diylbis oxy diethanamine, 3,6-dioxaoctamethylenediamine, 2-2-2-aminoethoxy ethoxy ethanamine, 3,6-dioxaoctane-1,8-diamine, ethylene glycol bis 2-aminoethyl ether
PubChem CID 70248
IUPAC Name 2-[2-(2-aminoethoxy)ethoxy]ethanamine
SMILES C(COCCOCCN)N

Specifications

Density 1.01 g/mL
Boiling Point 105°C to 109°C (6 mmHg)
Flash Point 121°C (250°F)
Refractive Index 1.461
Linear Formula C6H16N2O2
Quantity 250 g
UN Number UN2735
Beilstein 1739187
Sensitivity Hygroscopic
Solubility Information Soluble in 0.4 parts water.
Formula Weight 148.2
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2,2'- (Ethylenedioxy)bis (ethylamine)
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 213-203-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.