Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. p-7137005 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 53001-71-1
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD01631453
InChI Key WTKVWOHBTGWNEO-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
PubChem CID 2734024
IUPAC Name 1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
SMILES C1=C(C(=C(C(=C1F)F)F)F)CBr

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
UN Number 3265
Formula Weight 243.00
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzyl Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.