Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. D50855G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. D50855G

Supplier: TCI America D50855G

Chemical Identifiers

CAS 401797-00-0
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD12405519
InChI Key HWIMTTLCFONAAB-UHFFFAOYSA-N
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene, 3,4-Dimethylphenylboronic Acid Pinacol Ester
PubChem CID 15906194
IUPAC Name 2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C)C

Specifications

Color White
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Formula Weight 232.13
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.