Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T111625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T111625G 25 g
T11165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T111625G Supplier TCI America Supplier No. T111625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 573-11-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002433
InChI Key HZNQSWJZTWOTKM-UHFFFAOYSA-N
Synonym benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid
PubChem CID 11308
IUPAC Name 2,3,4-trimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)C(=O)O)OC)OC

Specifications

Melting Point 100°C
Color White
Quantity 25 g
Formula Weight 212.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trimethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.