Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trimethylpentane, 98+%

Catalog No. p-7044776
Click to view available options
Quantity:
5 g
25 g

CAS: 565-75-3 | C8H18 | 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-75-3
Molecular Formula C8H18
MDL Number MFCD00008924
InChI Key RLPGDEORIPLBNF-UHFFFAOYSA-N
PubChem CID 11269
IUPAC Name 2,3,4-trimethylpentane
SMILES CC(C)C(C)C(C)C

Specifications

Melting Point -110°C
Density 0.718
Boiling Point 113°C to 115°C
Flash Point 5°C (41°F)
Refractive Index 1.404
Quantity 5 g
UN Number UN1262
Beilstein 1696869
Synonym pentane, 2,3,4-trimethyl, unii-woj1qvr6oa, woj1qvr6oa, 2,3,4-trimethyl-pentane, pentane,3,4-trimethyl, acmc-1ao24, pentane,2,3,4-trimethyl, 4-01-00-00446 beilstein handbook reference, 2,3,4-trimethylpentan-3-yl, 2,3,4-trimethylpentane
Molecular Weight (g/mol) 114.232
Formula Weight 114.23
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethylpentane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 209-292-6
RTECSNumber SA3321500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.