Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',5'-Tri-O-acetyluridine 98.0+%, TCI America™
SDP

Catalog No. T254925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4105-38-8
Molecular Formula C15H18N2O9
Molecular Weight (g/mol) 370.31
MDL Number MFCD00023795
InChI Key AUFUWRKPQLGTGF-FMKGYKFTSA-N
Synonym Uridine 2′C,3′C,5′C-Triacetate
PubChem CID 124300592
IUPAC Name [(2R,3R,4R,5R)-3,4-bis(acetyloxy)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(C)=O)[C@@H]1OC(C)=O)N1C=CC(=O)NC1=O

Specifications

Melting Point 129°C
Color White
Quantity 25 g
Formula Weight 370.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,5′-Tri-O-acetyluridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.