Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chlorophenyl)ethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13078-79-0
C8H10ClN
155.63
MFCD00047957
NRHVNPYOTNGECT-UHFFFAOYSA-N
2-3-chlorophenyl ethylamine,2-3-chlorophenyl ethanamine,3-chlorophenethylamine,2-3-chlorophenyl ethan-1-amine,2-3-chloro-phenyl-ethylamine,3-chlorophenylethanamine,3'-chlorophenethylamine,m-chlorophenethylamine,2-m-chlorophenyl ethylamine,benzeneethanamine, 3-chloro
139381
2-(3-chlorophenyl)ethanamine
C1=CC(=CC(=C1)Cl)CCN
13078-79-0
C8H10ClN
155.63
MFCD00047957
NRHVNPYOTNGECT-UHFFFAOYSA-N
2-3-chlorophenyl ethylamine,2-3-chlorophenyl ethanamine,3-chlorophenethylamine,2-3-chlorophenyl ethan-1-amine,2-3-chloro-phenyl-ethylamine,3-chlorophenylethanamine,3'-chlorophenethylamine,m-chlorophenethylamine,2-m-chlorophenyl ethylamine,benzeneethanamine, 3-chloro
139381
2-(3-chlorophenyl)ethanamine
C1=CC(=CC(=C1)Cl)CCN
Boiling Point 111.0°C to 113.0°C (12.0 mmHg)
Color Yellow
Flash Point >109°C
Density 1.1190g/mL
Specific Gravity 1.119
Infrared Spectrum Authentic
Quantity 5g
Refractive Index 1.5480 to 1.5500
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2N(CH2)2C6H4Cl
Formula Weight 155.63
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-(3-Chlorophenyl)ethylamine, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA