Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-Dideoxycytidine, 98%

Catalog No. AC449505000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449505000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449505000 Supplier Thermo Scientific Chemicals Supplier No. 449505000

CAS: 7481-89-2 | C9H13N3O3 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7481-89-2
Molecular Formula C9H13N3O3
Molecular Weight (g/mol) 211.22
MDL Number MFCD00012188
InChI Key WREGKURFCTUGRC-KGQMAECUNA-N
Synonym zalcitabine, 2',3'-dideoxycytidine, dideoxycytidine, ddcyd, hivid, ddc, cytidine, 2',3'-dideoxy, zalcitibine, ddc antiviral, 4-amino-1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one
PubChem CID 24066
ChEBI CHEBI:10101
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1CC[C@@H](CO)O1

Specifications

Melting Point 216.0°C to 220.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Merck Index 1510307
Solubility Information Solubility in water: soluble
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material 2', 3'-Dideoxycytidine
Hazard Category Carcinogenicity (category 2)
Hazard Statement Suspected of causing cancer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.