Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorobenzylamine, 97%, Thermo Scientific™

Catalog No. p-4228602
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72235-51-9
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00070793
InChI Key OHZUCDHZOHSBPZ-UHFFFAOYSA-N
Synonym 2,3-difluorobenzylamine, 2,3-difluorophenyl methanamine, 2,3-difluorobenzyl amine, 2,3-difluorobenzyamine, benzenemethanamine, 2,3-difluoro, 2,3-difluoro-benzylamine, 2,3-difluorophenyl methylamine, 1-2,3-difluorophenyl methanamine, pubchem7356, 2,3-difluorophenyl methamine
PubChem CID 2736941
IUPAC Name (2,3-difluorophenyl)methanamine
SMILES C1=CC(=C(C(=C1)F)F)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2230g/mL
Boiling Point 65°C (4.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.223
Formula Weight 143.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Difluorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.