Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzoic acid, 99%

Catalog No. p-3757727
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle

CAS: 1521-38-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1521-38-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002432
InChI Key FODBVCSYJKNBLO-UHFFFAOYSA-N
Synonym o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
PubChem CID 15204
IUPAC Name 2,3-dimethoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121°C to 124°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CO2H
Quantity 5 g
Beilstein 10, I, 174
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 182.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 3-Dimethoxybenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 216-188-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.