Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbutane, 98+%

Catalog No. p-3778371
Click to view available options
Quantity:
5 mL
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 79-29-8 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -129.0°C
Color Colorless
Density 0.6620g/mL
Boiling Point 58.0°C
Flash Point −29°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.3740 to 1.3760
Linear Formula (CH3)2CHCH(CH3)2
Quantity 5 mL
Beilstein 01,151
Specific Gravity 0.662
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 86.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethylbutane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-193-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.