Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hydroxide, 95%, pure

Catalog No. AC315720250
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC315720250

Supplier: Thermo Scientific Chemicals 315720250

Only null left
Add to Cart

CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 94% min. (Complexometry)
Linear Formula Mg(OH)2
Merck Index 15, 5734
Solubility Information Solubility in water: insoluble
IUPAC Name magnesium;dihydroxide
Formula Weight 58.33
Percent Purity 95%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium hydroxide, 95%
Show More Show Less
Hazard Category AIR SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-170-3
RTECSNumber OM3570000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.