Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylindole 99%, Thermo Scientific™

Catalog No. AC153080250
Click to view available options
Quantity:
5 g
25 g

Catalog No. AC153080250

Supplier: Thermo Scientific Chemicals 153080250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-55-4
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
InChI Key PYFVEIDRTLBMHG-UHFFFAOYSA-N
Synonym 2,3-dimethylindole, 1h-indole, 2,3-dimethyl, indole, 2,3-dimethyl, 2,3-dimethyl indole, tfw7o9hwzk, unii-tfw7o9hwzk, 2,3-dimethyl-1h-indol, 2,3-dimethyl-indol, 2,3-dimethyl-indole, pubchem7334
PubChem CID 7053
IUPAC Name 2,3-dimethyl-1H-indole
SMILES CC1=C(NC2=CC=CC=C12)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104°C to 107°C
Color White to Yellow
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 20, 319
Formula Weight 145.2
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 2, 3-Dimethylindole, 99%
Show More Show Less
EINECSNumber 202-076-2
RTECSNumber NL7185000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.