Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylpentane, 97%

Catalog No. AA4370714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4370714 25 g
AA4370706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4370714 Supplier Thermo Scientific Chemicals Supplier No. 04370714
Only null left
Add to Cart

CAS: 565-59-3 | C7H16 | 100.21 g/mol

It is used in the liquid-phase oxidation of 2,4-dimethylpentane and in a study on the preparation and the physical constants of a number of alkanes and cycloalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-59-3
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009337
InChI Key WGECXQBGLLYSFP-UHFFFAOYNA-N
Synonym pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
PubChem CID 11260
IUPAC Name 2,3-dimethylpentane
SMILES CCC(C)C(C)C

Specifications

Density 0.690 g/mL
Boiling Point 89°C to 90°C
Flash Point −6°C (21°F)
Refractive Index 1.392
Quantity 25 g
UN Number UN1206
Solubility Information Insoluble in water.
Formula Weight 100.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylpentane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber 209-280-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.