Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-Isopropylideneuridine

Catalog No. p-7043958
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 362-43-6 | C12H16N2O6 | 284.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362-43-6
Molecular Formula C12H16N2O6
Molecular Weight (g/mol) 284.268
MDL Number MFCD00034509
InChI Key GFDUSNQQMOENLR-PEBGCTIMSA-N
Synonym 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 2/',3/'-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 64967
IUPAC Name 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione
SMILES CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C

Specifications

Melting Point 165°C to 166°C
Quantity 1 g
Beilstein 32671
Formula Weight 284.27
Chemical Name or Material 2',3'-Isopropylideneuridine
EINECSNumber 206-647-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.