Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Lutidine, 98+%

Catalog No. AC268765000
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Catalog No. AC268765000

Supplier: Thermo Scientific Chemicals 268765000

CAS: 583-61-9 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-61-9
MDL Number MFCD00009605
InChI Key HPYNZHMRTTWQTB-UHFFFAOYSA-N
Synonym 2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
PubChem CID 11420
IUPAC Name 2,3-dimethylpyridine
SMILES CC1=C(N=CC=C1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -17.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 161.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 0.7% max. 2-Ethylpiridine (GC)
Packaging Glass bottle
Molecular Formula C7H9N
Refractive Index 1.5062 to 1.5082
Quantity 500 mL
Beilstein 20, 243
Specific Gravity 0.95
Solubility Information Solubility in water: 95g/L (26°C). Other solubilities: soluble in methanol and ether,miscible with dmf and thf
Molecular Weight (g/mol) 107.16
Formula Weight 107.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2, 3-Lutidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-514-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.